Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylindoline

Supplier: APOLLO SCIENTIFIC
Warning

OR10176-10G OR10176-100G
76561-294EA 31 USD InStock
76561-294 76561-354
2-Methylindoline
2-Methylindoline


Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 228…229 °C
Melting Pt: –51 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty