Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinopyrimidine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Hydrazinylpyrimidine

OR302061-1G OR302061-5G OR302061-25G
76573-928EA 114 USD InStock
76573-928 76573-924 76573-926
2-Hydrazinopyrimidine 97%
2-Hydrazinopyrimidine


Formula: C₄H₆N₄
MW: 110.12 g/mol
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD01249337
CAS Number: 7504-94-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty