Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Octylphosphonic acid 97%

Supplier: APOLLO SCIENTIFIC
Synonyms: OPA1-Octylphosphonic acidn-Octylphosphonic acid

OR922324-25G OR922324-100G
77411-798EA 228 USD InStock
77411-798 77411-800
Octylphosphonic acid 97%
Octylphosphonic acid


Formula: C₈H₁₉O₃P
MW: 194.21 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00015841
CAS Number: 4724-48-5
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty