Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Chlorophenyl)piperidine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Chloro-4-(piperidin-2-yl)benzene hydrochloride

OR1006-1G OR1006-100MG OR1006-250MG
77402-432EA 140 USD InStock
77402-432 77402-442 77402-440
2-(4-Chlorophenyl)piperidine hydrochloride
2-(4-Chlorophenyl)piperidine hydrochloride


Formula: C₁₁H₁₄ClN·ClH
MW: 232.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02178960
CAS Number: 1172776-65-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty