Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC2076G-250MG PC2076G-1G PC2076G-5G
77427-516EA 147 USD InStock
77427-516 77427-514 77427-512
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine 95%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)piperazine


Formula: C₁₀H₁₁ClF₃N₃
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00173934
CAS Number: 132834-59-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty