Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodo-5-(trifluoromethyl)aniline 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Amino-4-iodobenzotrifluoride

PC2015-1G PC2015-10G PC2015-5G
77427-488EA 101 USD InStock
77427-488 77427-486 77427-484
2-Iodo-5-(trifluoromethyl)aniline 98%
2-Iodo-5-(trifluoromethyl)aniline


Formula: C₇H₅F₃IN
MW: 287.0210 g/mol
Boiling Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD04972658
CAS Number: 105202-02-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty