Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlor-4-fluorothiophenol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Chloro-3-fluoro-6-mercaptobenzene2-Chloro-4-fluorobenzenethiol2-Chlor-4-fluorobenzenethiol2-Chlor-4-fluorobenzene-1-thiol

PC1889-5G PC1889-250MG PC1889-1G
77427-358EA 54 USD InStock
77427-358 77427-362 77427-360
2-Chlor-4-fluorothiophenol 97%
2-Chlor-4-fluorothiophenol


Formula: C₆H₄ClFS
MW: 162.61 g/mol
Boiling Pt: 86…88 °C/15 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00052936
CAS Number: 175277-99-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty