Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-5-fluoro-4-methylpyridine 95%

Supplier: APOLLO SCIENTIFIC

PC201059-250MG PC201059-1G PC201059-5G
77427-448EA 313 USD InStock
77427-448 77427-446 77427-444
3-Bromo-5-fluoro-4-methylpyridine 95%
3-Bromo-5-fluoro-4-methylpyridine


Formula: C₆H₅BrFN
MW: 190.0130 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257616
CAS Number: 1211517-76-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty