Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-fluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1880G-5G PC1880G-100G PC1880G-25G
77427-354EA 13 USD InStock
77427-354 77427-352 77427-350
4-Chloro-3-fluorophenol 98%
4-Chloro-3-fluorophenol


Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 84 °C/44 mmHg
Melting Pt: 50…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042583
CAS Number: 348-60-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty