Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2554H-25G PC2554H-5G PC2554H-1G
77427-700EA 77 USD InStock
77427-700 77427-698 77427-696
2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine 97%
2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine


Formula: C₇H₅Cl₂F₃N₂
MW: 245.03 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085001
CAS Number: 86398-94-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty