Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluoro-4-nitrobenzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4399-100G PC4399-5G PC4399-25G
77429-724EA 32 USD InStock
77429-724 77429-722 77429-726
3-Fluoro-4-nitrobenzoic acid 97%
3-Fluoro-4-nitrobenzoic acid


Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 173…176 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862092
CAS Number: 403-21-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty