Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-(trifluoromethyl)aniline

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Amino-3-chloro-α,α,α-trifluortoluene2-Amino-3-chloro-α,α,α-trifluorotoluene6-Chloro-α,α,α-trifluoro-o-toluidine

PC407008-25G PC407008-10G
77429-354EA 40 USD InStock
77429-354 77429-352
2-Chloro-6-(trifluoromethyl)aniline
2-Chloro-6-(trifluoromethyl)aniline


Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272565
CAS Number: 433-94-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty