Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethyl)benzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-(Hydroxymethyl)benzal fluoride[3-(Difluoromethyl)phenyl]methanol

PC49199-5G PC49199-1G
77430-390EA 51 USD InStock
77430-390 77430-392
3-(Difluoromethyl)benzyl alcohol 97%
3-(Difluoromethyl)benzyl alcohol


Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19053257
CAS Number: 1309875-03-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty