Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2389-1G PC2389-5G PC2389-10G
77427-638EA 13 USD InStock
77427-638 77427-636 77427-640
4-Methoxy-3-(trifluoromethyl)benzaldehyde 98%
4-Methoxy-3-(trifluoromethyl)benzaldehyde


Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD01091012
CAS Number: 50823-87-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty