Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-(pentafluorothio)aniline

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Amino-3-chlorophenylsulphur pentafluoride

PC405667-5G PC405667-10G PC405667-1G
77429-334EA 1645 USD InStock
77429-334 77429-338 77429-336
2-Chloro-4-(pentafluorothio)aniline
2-Chloro-4-(pentafluorothio)aniline


Formula: C₆H₅ClF₅NS
Melting Pt: 56…58 °C
Storage Temperature: Refrigerator
CAS Number: 165114-85-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty