Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Boc-Cis-4-F-Pro-OH(2S,4S)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acidN-(tert-Butoxycarbonyl)-cis-4-fluoro-L-proline(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acidN-Boc-cis-4-fluoro-L-proline(2S,4S)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid

PC6794-100G PC6794-5G
77432-780EA 17 USD InStock
77432-780 77432-776
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid


Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
Melting Pt: 157…162 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973957
CAS Number: 203866-13-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty