Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide

Supplier: APOLLO SCIENTIFIC
Warning

PC450563-250MG PC450563-1G
77429-976EA 722 USD InStock
77429-976 77429-978
N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide
N-Boc-2,3,4,5,6-pentafluorobenzene-1-carboximidamide


Formula: C₁₂H₁₁F₅N₂O₂
MW: 310.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30062918
CAS Number: 1980048-36-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty