Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-2,5-Difluorocinnamic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (E)-3-(2,5-Difluorophenyl)acrylic acid

PC2762-5G PC2762-100G PC2762-25G
77427-820EA 17 USD InStock
77427-820 77427-818
trans-2,5-Difluorocinnamic acid 97%
trans-2,5-Difluorocinnamic acid


Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00010318
CAS Number: 112898-33-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty