Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
KODAK® BioMax® Maximum Resolution (MR) Autoradiography Film, Carestream Health
KODAK® BioMax®
8941114 8572786 8952855 8701302
IB-IB8701302CS 4656.21 USD InStock
IB-IB8701302 IB-8952855 IB-8941114 IB-8572786
KODAK® BioMax® Maximum Resolution (MR) Autoradiography Film, Carestream Health
Autoradiography Films Films
A sensitive, clear, maximum resolution film recommended for detection of 35S, 33P, 14C
Suitable for chemiluminescence although not as sensitive as BioMax® Light film. Incorporates advanced KODAK® emulsion technology in a single emulsion format. Suitable for manual and automated processing.
Ordering information: MR-2 sheets have interleaved packaging (sheets are individually wrapped). MR-1 sheets have non-interleaved packaging.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty