Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC502497-1G PC502497-5G
77430-884EA 502 USD InStock
77430-884 77430-886
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde


Formula: C₈H₃F₅O₂
MW: 226,1 g/mol
MDL Number: MFCD31561166
CAS Number: 2244081-97-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty