Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Mini Thermo-Anemometers, Flir Commercial Systems, INC
Supplier: Flir
45118 45158
89030-358EA 268.75 USD InStock
89030-358 89030-360
Mini Thermo-Anemometers, Flir Commercial Systems, INC
Anemometers
Meters measure and display air velocity and temperature
Meter 89030-360 also features a dual display of air velocity and relative humidity with dew point.
Air velocity can be measured in fpm, mph, m/s, km/h, knots, or Beaufort force, with a selectable averaging function of 5, 10, or 13-second intervals. Data hold function freezes most recent display and auto shutoff initiates 20 minutes after the last key is pressed. The water-resistant housing floats and has been drop-tested to 1.8m (6'). Plastic impeller is noncorrosive and replaceable. Meter housings can fold up or extend to 22.9cm (9").
Certifications: CE marked.
Ordering information: Meters include CR2032 lithium battery and lanyard.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty