Would you like to log in again?
Static-Dissipative Binders, SECO
Supplier: SECO
BI-3X BI-1X BI-1.5X BI-2X
58605-946CS 660.43 USD InStock
58605-946 58605-958 58605-956 58605-960
Static-Dissipative Binders, SECO
Binders and Indexes
Three-ring, static-dissipative vinyl binders are made using stabilized PVC-based film
Designed for use at approved field force workstations and static-sensitive environments. Binders feature heavy-duty, nickel-plated steel rings; 20-gauge vinyl sealed over heavy chipboard; durable easy-open and -close triggers; and reinforced tear-resistant hinge seams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty