Would you like to log in again?
VWR® Compact Sling Psychrometer
Supplier: VWR
VWR®
61503-0111
61222-744EA 101.32 USD InStock
61222-744
VWR® Compact Sling Psychrometer
Hygrometers Psychrometers
Measures relative humidity with weather bureau-type wet and dry bulb.
  • Organic liquid
  • Pocket-sized plastic case
Organic liquid-filled thermometer with white back glass are mounted in a pocket-sized self contained plastic case designed with a built-in slide rule to convert wet and dry bulb readings to relative humidity, thus eliminating the need for calculating tables.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty