Would you like to log in again?
Coliform Water Test Sample Container, Capitol Vial®
Supplier: Capitol Vial
04HP83HCNP 04HP83HCPL
89049-008CS 150.78 USD InStock
89049-008 35000-892
Coliform Water Test Sample Container, Capitol Vial®
Vials
Pure polypropylene (sterile-by-process environment) container used to test for coliform.
  • Locking arrow assures sterility
  • With or without Sodium thiosulfate tablet
One-piece construction with leak-tight, attached lid and custody seal reduces contamination.
Int.Ø×H: 43×95 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty