Would you like to log in again?
Clorox® Healthcare® Citrace® Hospital Disinfectant and Deodorizer
Supplier: Essendant
CLO49100
21850-242CS 249.37 USD InStock
21850-242
Clorox® Healthcare® Citrace® Hospital Disinfectant and Deodorizer
Cleaning Agents Disinfectants
Clorox Healthcare® Citrace® hospital disinfectant and deodorizer kill microorganisms on soft surfaces, hard surfaces and more.
  • Fresh citrus scent fades quickly after spraying
  • Eliminates odors instantly by killing the bacteria that cause them
  • Prevents mold and mildew on fabrics
  • Intended for use in commercial settings
  • Liquid form in aerosol can
It kills 99.9% of bacteria on soft surfaces in just 30 seconds. It kills 24 microorganisms on hard surfaces in five minutes. With a fresh citrus scent, Citrace® eliminates odors from the air instantly. Effective against bacteria, fungus, germs, mildew, mold, viruses. For use on ceramic, cement, fiberglass, glass, grout, laminates, metal, plastic, vinyl.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty