Would you like to log in again?
Ambidextrous Wrist Supports, OK-1® Safety, OccuNomix
Supplier: OccuNomix
OK-1® Safety
AMBI L
39870-302CS 389.51 USD InStock
39870-302
Ambidextrous Wrist Supports, OK-1® Safety, OccuNomix
Ergonomic Supports Wrist and Hand Supports
Designed to be used on either the left hand or right hand, supports provide the ultimate in comfort and practicality
Constructed of sturdy fabric-lined neoprene. Utilize an 8cm wide wrap-around strap for added compression, along with flexible inserts that help limit hand flexion and extension.
Ordering information: Measure wrist circumference and order applicable size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty