Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VWR® Signature™ Inoculating Loops and Needles, Sterile
Supplier: Avantor
VWR® Signature™
175CS50T.VWR 177CS50T.VWR 179CS50T.VWR 176CS50T.VWR 178CS50T.VWR
30002-096CS 519.29 USD InStock
30002-096 30002-098 30002-092 30002-094 30002-090
VWR® Signature™ Inoculating Loops and Needles, Sterile
Spreaders Inoculating Loops and Needles
Calibrated disposable polystyrene inoculation loops and needles.
  • Disposable polystyrene fabrication
  • Color-coded for easy size identification
  • Available in soft or rigid plastic
  • Ultra-smooth surface for easy streaking
Loops are available in soft or rigid plastic and feature an ultra smooth surface for easy streaking. Loops and needles are color-coded for easy size identification.
Packaging: Packaged in anti-roll tubes with push-on resealable cap, 50 loops/needles per tube, 20 tubes per case. Length: 21 cm (81/4").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty