Would you like to log in again?
SMITH & WESSON® Code 4™ Safety/Shooting Glasses, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Smith & Wesson®
19832 19834 19836 19835 19833
67200-224EA 27 USD InStock
67200-224 67200-226 14227-562 14227-560 67200-222
SMITH & WESSON® Code 4™ Safety/Shooting Glasses, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Lightweight, stylish safety glasses weigh less than one ounce and feature shatter-resistant polycarbonate wraparound lenses for excellent peripheral vision.
  • Hinge stops prevent scratched lenses
  • Contoured temple tips
  • Ribbed, nonslip rubber nosepiece
  • 99.9% UV protection
All glasses feature black frames, with the exception of model 67200-222, which have platinum frames.
Certifications: ANSI Z87.1
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty