Would you like to log in again?
Calbiochem® Caspase-1 Inhibitor I
Supplier: EMD Millipore
Calbiochem®
400010-1MG 400010-5MG 400010-5MG 400010-1MG
80052-266EA 101.27 USD InStock
80052-266 CA80052-268 CA80052-266 80052-268
Calbiochem® Caspase-1 Inhibitor I
Proteins and Peptides
A potent, specific, and reversible inhibitor of caspase-1 (Ki=200pM for human recombinant caspase-1) and caspase-4
Strongly inhibits anti-APO-1 induced apoptosis in L929-APO-1 cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty