Would you like to log in again?
dNTPs, Omega Bio-Tek
Supplier: Omega Bio-Tek
TQAC135 TQAC135
101414-958EA 182.11 USD InStock
CA101414-958 101414-958
dNTPs, Omega Bio-Tek
Nucleic Acid Reagents Nucleotides
Each ultrapure 2'-deoxynucleotide 5'-triphosphate (dATP, dCTP, dGTP, dTTP) is diluted in a stabilized solution (pH 7.5)
They are ready to use for PCR or other reactions. Each dNTP is tested at 99% purity by HPLC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty