Would you like to log in again?
HEPA Filtered Lead Vacuum, Minuteman®
Supplier: Minuteman
C82985-06
62510-088EA 1590.51 USD InStock
62510-088
HEPA Filtered Lead Vacuum, Minuteman®
Vacuum Cleaners
Lead vacuum is designed for safe, efficient daily use in commercial and industrial applications to recover lead, dust, and paint chips
Features a durable polyethylene tank, acoustically-designed lid to minimize noise, and a “sure grip/quick-release” mechanism for easy connection and removal of hoses. Vacuum contains particulates within a special filter, allowing no hazardous dust to escape. Special interlocking safety system prevents vacuum use without proper alignment of HEPA filter. HEPA filter is 99.99% efficient at 0.12µ.
Certifications: UL approved.
Ordering information: Comes complete with tank, 50' cord and dry tool kit, consisting of a 304.8 cm (10') hose, wand, adapter, dusting brush, 30.5 cm (12") crevice tool and a 35.6 cm (14") floor brush.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty