Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Safety Shields, Mitchell Plastics™
Supplier: Mitchell Plastics
RP-100 RP-650 RP-050 SS-100 RP-750 RP-150 RP-025 RP-145 RP-100C RP-850 RP-800 RP-125C RP-700 RP-852
11380-795EA 540.66 USD InStock
11380-795 25405-348 25405-328 25405-330 25405-342 25405-332 25405-356 25405-334 56615-963 56615-964 56615-965 56615-966 56615-868 56615-968
Safety Shields, Mitchell Plastics™
Shields Safety Shields
Acrylic shields provide a protective barrier between lab personnel and biohazardous/beta materials
  • Each shield tapers back at an angle for easy viewing and protection
  • A variety of shield styles and designs accommodate varied workplace requirements
  • Different sizes of balance shields are also available for use with Mettler, Sartorius, and general balances
Stand alone, freestanding Biohazard Shields taper back at an angle for easy viewing. This design features a deep base, which offers a large work area for test tube racks, pipets, and other accessories. The Slim-Line shield is a narrower version designed to allow easier access to the work site. The smallest cell-counter shield is adjustable and may be used with small manual or semi-automated hematology counters. Its base slides under the counter for added stability. These acrylic, adjustable shields may be mounted with feet and adjustable arms to a counter or attached to equipment such as a hematotology counter with double sided tape. Benchtop tapered shields provide more arm flexibilty and increased arm movement, reducing arm fatigue. The lower portion is only 30.5cm (12") wide and the top portion of the shield is 50.8cm (20") wide to provide the maximum barrier between personnel and hazardous materials. Shields are tapered back at an angle for easy viewing. Adjustable Beta shields are clear and can be adjusted 20° from a vertical position to accommodate space requirements. Two stabilizers support the shield on both sides to prevent tipping. The extra large sizes are especially good for use on the floor. Flex shields are mounted to 45.7cm (18") flexible tubes to position the shield. The 10.2x10.2cm (4x4") base of each flexible tube can be mounted to a counter or a wall with screws or double sided tape. The Beta Flex shield is supported by two flexible tubes to hold the shield in desired position. The Septa shield is a protective shield with base that is worn around the neck and held in place with a soft, adjustable Velcro® strap. It is lightweight (9oz.) and protects the face and neck from splashes, sprays, and abrasions. Balance Shields are available for Mettler and Sartorius balances to protect samples during weighing. They enclose the balance, leaving controls exposed for easy access. The hinged lid offers direct access to the weighing pan. A 5.1cm (2") hole on the top of the lid allows for sample dispensing. The standard large and small balance shields are designed to protect sensitive measurings from drafts, while maintaining a controlled environment. A removable lid allows additional access when pouring liquids or adding dry chemicals from the top. Both shields have three sides, each with a notch at the bottom to position a power cord where needed.
Deep base shields provide a large work area for test tube racks, pipets, and other accessories. Slim-line profile shields enable the technician to reach around the shield with both hands for easy access to the work site. The cell counter shield fits on small manual and semi-automatic hematology counters, with a base that slides under the counter for added stability. Small shields are available with adjustable arms that attach to counter tops or hematology counters to provide protection from blood aerosols. Tapered biohazard/beta shields feature a curved lower portion and wide top to allow arm flexibility and increased movement. Large shields are adjustable from their vertical positions for protection. The Face Protector is a reusable shield with base that is comfortably worn around the neck and held in place by a soft, velcro adjustable strap.
Accessories information: An accessory base, C-Clamps, and magnetic holders are optional accessories available for use with Flex Shields.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty