Would you like to log in again?
VWR® Dry Mop Heads
Supplier: VWR
VWR®
MZDM62-14 MZDM61-14 MZDM61-18
68400-820CS 584.03 USD InStock
68400-820 68400-798 68400-796
VWR® Dry Mop Heads
Mops
Mops absorb detergent quickly and may be used on floors, walls, and ceilings without a bucket or wringer.
  • Latex/rayon blend mop head
  • May be used with disinfectants
  • Autoclavable
Material is designed for applications in which excess moisture is a concern. A special fabric strip allows for fast-drying action. Mop heads are sturdy enough to be laundered and reused. Latex/rayon blend mop head offers absorbency similar to that of PVA, but may be used with disinfectants and are autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty