Would you like to log in again?
VWR® Standard Standard Hotplates
Supplier: VWR
VWR®
984VW7AHPUSS 984VW0CHPUSS 984VW4AHPUSS 984VW7CHPUSS 984VW0AHPUSS 984VW4CHPUSS
97042-610EA 499.93 USD InStock
97042-610 97042-654 97042-614 97042-570 97042-650 97042-574
VWR® Standard Standard Hotplates
Heaters Hot Plates
Standard hotplates are designed for general-purpose laboratory use.
  • Excellent temperature uniformity
  • Enhanced electronics
  • Cool touch, chemical-resistant housing
  • Best-in-Class Five-Year Warranty
  • Industry Leading Safety Features
These analog units are microprocessor controlled and equipped with easy-to-use, dial-adjustment controls
Hotplates are available with ceramic or aluminum top plates. Ceramic top plates feature an easy-to-clean, chemical-resistant, reflective white surface. Aluminum top plates will not crack or chip, and offer a more even heating surface. The unit's housing is constructed of a heat-resistant polymer that remains cool to the touch, and is chemical-resistant as well. For additional safety, a hot symbol warning light is illuminated while the unit is in use, and remains lit until the top plate is sufficiently cooled.Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
Enhanced electronics and a robust heater regulate the heating surface and allow units to reach the desired set point quickly. The low-profile design requires minimal bench space and permits use within a fume hood. The spill-resistant housing channels fluids away from internal components in the event of a spill.
Certifications: RoHS certified. TUV listed. CE marked.
Ordering information: Units are supplied with a 234 cm (92") detachable, 3-wire cord and plug. The optional support rod and clamp kit includes a 45.7 cm (18") long stainless steel support rod, a thermometer/temperature probe extension clamp, a three-prong dual-adjust swivel clamp, and a hook connector. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty