Would you like to log in again?
SP Wilmad-LabGlass U-Shaped Drying Tubes, SP Industries
Supplier: SP Industries
LG-6790-102
14210-172EA 145.34 USD InStock
14210-172
SP Wilmad-LabGlass U-Shaped Drying Tubes, SP Industries
Tubes Drying Tubes
These U-shaped borosilicate glass drying tubes have a Standard Taper joint at one end and are designed to avoid desiccant contact with reaction.
The other end is open to receive a size #2 rubber stopper with bent inlet tube (included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty