Would you like to log in again?
Amersham™ Hybond™-N+ Western Blotting Membrane, Cytiva
Supplier: Cytiva
Amersham™ Hybond™
RPN203B RPN1210B RPN119B RPN82B RPN1520B RPN1210B RPN1520B RPN119B RPN2020B RPN203B RPN1510B RPN1510B RPN2020B RPN303B RPN82B RPN303B
95038-376EA 658.56 USD InStock
CA95038-376L CA95038-342L CA95038-362L CA95038-336L CA95038-424L CA95038-302L CA95038-400L 95038-376 95038-342 95038-362 CA95038-308L 95038-308 95038-424 95038-336 95038-302 95038-400
Amersham™ Hybond™-N+ Western Blotting Membrane, Cytiva
Transfer Membranes
Amersham™ Hybond™-N+ is a positively charged nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm² recommended for use with radioactive or non radioactive chemiluminescence and chemifluorescence detection systems.
  • Recommended for all high density membrane arrays
  • Multiple reprobing is possible without loss of membrane integrity
  • QC tested in genomic Southern blots
Amersham™ Hybond™-N+ yields excellent sensitivity in conventional Southern blotting. Nucleic acid samples may be fixed by UV exposure or baking, although UV fixation is best for maximum reproducibility.
Caution: Amersham™ Hybond™-N+ is not suitable for alkali blotting and fixation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty