Would you like to log in again?
High-Purity Polypropylene Faucets, Scientific Plastics
Supplier: Scientific Plastics
SPF-40STD SPF-30STD SPRF-1 SPF-40VB SPF-20VB SPF-10STD SPF-10VB SPF-30VB SPF-20STD
89013-982EA 2660.24 USD InStock
89013-982 89013-992 89013-980 89013-990 89013-988 89013-986 89013-996 89013-984 89013-994
High-Purity Polypropylene Faucets, Scientific Plastics
Faucets and Sinks Faucets
Ideal for the transportation of deionized water, distilled water, or other liquids where a high degree of purity is required
Faucets feature excellent chemical resistance and are made from pure unpigmented virgin Type I homopolymer, using no anti-oxidants, pigments, metallics, or other foreign substances. The smooth, nonporous polypropylene surfaces do not attract contaminants or provide breeding places for bacteria and other biological impurities. Spread gooseneck measures 15.2 cm (6"). Maximum temperature: 99 °C (210 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty