Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PD-1/PD-L1 Inhibitor 1 ≥99.50%

Supplier: SELLECK CHEMICALS LLC MS
Synonyms: (S)-1-(2,6-Dimethoxy-4-((2-methyl-[1,1'-biphenyl]-3-yl)methoxy)benzyl)piperidine-2-carboxylic acidBMS-1(2S)-1-[[2,6-dimethoxy-4-[(2-methyl[1,1'-biphenyl]-3-yl)methoxy]phenyl]methyl]-2-piperidinecarboxylic acid

S7911-5MG
103547-356EA 192.31 USD InStock
103547-356
PD-1/PD-L1 Inhibitor 1 ≥99.50%
PD-1/PD-L1 Inhibitor 1
A small-molecule inhibitor of PD-1/PD-L1 interaction with IC50 of 6 nM


Formula: C₂₉H₃₃NO₅
MW: 475.58 g/mol
MDL Number: MFCD28978741
CAS Number: 1675201-83-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty