Would you like to log in again?
ET8 Universal Input Chart Recorder, Dickson®
Supplier: Dickson
ET855
14905-002EA 1530.36 USD InStock
14905-002
ET8 Universal Input Chart Recorder, Dickson®
Chart Recorders Recorders
Designed for a wide variety of applications, this recorder features high/low audible alarm, user calibration, large display, user selectable output ranges and recording times, and two input channels
The high/low alarm displays minimum/maximum readings with no controller/relay functions. Unit uses 20.3cm (8") charts and has remote sensing capability with powered voltage or current transmitters. Two screw terminals on the back of the unit allow for easy connection of input transmitter. Free-standing or wall-mountable.
Output/chart ranges: 000.0 to 100.0, 0000 to 1000, 00.00 to 14.00, 000.0 to 150.0, 000.0 to 200.0,000.0 to 300.0, –10.00 to 10.00, –150.0 to 250.0
Input ranges: 0–20 mA, 4–20 mA, 0–5 Vdc, 1–5 Vdc
Ordering information: Supplied with two pens (red and blue), 120V AC adapter, and instruction manual.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty