Would you like to log in again?
Precise™ Basic Glove Box, Labconco®
Supplier: Labconco
Precise™
5220020 5220000
89038-774EA 11024.53 USD InStock
89038-774 89038-772
Precise™ Basic Glove Box, Labconco®
Glove Boxes and Glove Bags
Designed to provide a leak-tight physical barrier between the user and the materials inside the glove box
Glove box meets Class 1 atmosphere containment conditions for oxygen leak rates per ISO 10648-2 test method. The 115V model is ETL listed with UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1 conformity. The 230V model is CE marked.
The unit features a dry powder, epoxy-coated steel superstructure, 0.6cm (1/4") thick laminated safety glass viewing window with powder-coated steel frame and neoprene gasket, and molded one-piece, medium-density, polyethylene shell. The cutout area is 67Wx31.8Hcm (2613/32x121/2") high, and the window can be easily removed for loading equipment. The space saving inner and outer transfer chamber doors pivot upward and are counterbalanced and equipped with quick latches. The interlocking shelf of the interior transfer chamber is made of dry powder, epoxy-coated steel. The black, chemical-resistant phenolic core work surface is sealed to the glove box floor. Unit has an interior ceiling-mounted electrical duplex receptacle with 5A maximum (100/115V model) or 2A maximum (230V model) circuit load, and includes switches for the light and innermost interior electrical receptacle. Glove box is factory leak tested with a mass spectrometer while pressurized with helium at 12.7cm (5") of water gauge. No detectable leaks greater than 1x10-4 mL/sec.
Ordering information: Supplied with pair of 76.2cm (30") long, 150mil thick neoprene gloves (size 93/4), and a 22W fluorescent lamp with easy replacement from the top. Glove box requires supporting base (not included). See 97002-494 series for supporting base and other accessories to help customize the unit to meet specific applications.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty