Would you like to log in again?
170 Series Digital Multimeters, Fluke®
Supplier: Fluke
1564560 1564549 1564551
26984-076EA 750.45 USD InStock
26984-076 26984-078 26984-080
170 Series Digital Multimeters, Fluke®
Electrical Meters Digital Multimeters
These versatile multimeters feature true-RMS AC voltage and current measurement for accurate reading on all waveforms
Models 177 and 179 feature backlit displays. The backlight function automatically turns off after two minutes to save battery life. Model 179 also measures temperature in °C and °F. Power-up options for Models 177 and 179 include disabling the backlight time out.
Meter is housed in a high-impact overmold case with built-in probe holder slots. An easy-access battery door allows battery change without breaking the calibration seal. The 6000 count digital display updates four times per second and the 33-segment analog bargraph display updates 40 times per second. Meters feature hold and auto-hold functions. The hold function freezes the display, and auto-hold function holds the present reading until it detects new stable input, then the meter beeps and displays the new reading. Measurement modes include minimum, maximum, and average. Power-up options include turning on all LCD segments, disabling the beeper, disabling the sleep mode, and enabling smoothing. Meters feature manual or auto-ranging functions. In auto-range the meter selects the range with the best resolution for the present measurement value. Meters also feature fast continuity/open detection as the beeper sounds with a stretched pulse for opens or shorts as brief as 250µs.
Certifications: CAT III rated for 1000V; CAT IV rated for 600V. CSA certified.
Ordering information: Manufacturer’s lifetime warranty. Meter can be calibrated without opening the case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty