Would you like to log in again?
Barnstead/Lab-Line High-Temperature Vacuum Ovens, Thermo Scientific
Supplier: Thermo Scientific
Barnstead/Lab-Line
3628A 3625A
89008-784EA 13315.47 USD InStock
89008-784 89008-816
Barnstead/Lab-Line High-Temperature Vacuum Ovens, Thermo Scientific
Ovens Vacuum Ovens
Ideal for elevated temperature aging tests, these ovens are designed for desiccation, vacuum embedding, plating, process control, and inert atmosphere applications
CSA approved.
Ovens feature a maximum temperature of 280 °C (536 °F). Radiant warm wall heaters are mounted outside of the chamber to conserve chamber working space and to eliminate the hazards of open wire heaters. Two fully adjustable chrome-plated shelves improve heat conduction to samples and are easily removed for cleaning. Vacuum lines, air lines, and oven interior are constructed of corrosion-resistant stainless steel for optimum chamber cleanliness and durability. Ovens feature a glass window in the door for clear sample viewing and a durable, powder-coated, heavy gauge steel exterior. Two LED's show oven conditions. Vacuum controls and relief valves are located on front of the unit for convenient monitoring. Compression fittings on the sidewall of the oven accept 0.6cm (1/4") hard tubing for pump and air connections. The built-in safety controller prevents temperature runaway with a back-up thermostat. Unit 89008-816 can accommodate two shelves and includes a single data point controller. Unit 89008-784 can accommodate up to eight shelves and features a seven-day programmable timer that cycles the oven on or off up to eight times per day.
Ordering information: Supplied with manufacturer's one-year warranty and two aluminum shelves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty