Would you like to log in again?
V-Safe™ Safety Lockouts for Valves, Honeywell Safety
Supplier: Honeywell Safety
V-Safe™
VS06R VS06B VS04R
13502-512CS 851.54 USD InStock
13502-512 13502-508 13502-504
V-Safe™ Safety Lockouts for Valves, Honeywell Safety
Security Locks
Prevent accidents by controlling unauthorized or accidental use of energy.
Valve cover is made of Polypro plastic and shaped into half-moons to completely cover the valve wheel. Lockout can be modified to fit smaller valves by drilling a hole aligned with the first and then closing the lockout to completely cover the valve. Cover is fully dielectric and resistant to cracking and abrasion. Plastic will not crack under extreme temperature changes, from –45.5 to 182°C (–50 to 360°F).
Red valves should be used to indicate fire-quenching materials such as halon. Yellow valves indicate materials that are inherently hazardous, such as propane gas. Blue valves are used for cases of inherently low hazard, such as argon. Green valves should be used to indicate liquids of inherently low hazard, such as mill water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty