Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® Laser Cartridge, 7553A, 7553MICR, 7553X, Essendant LLC MS

Supplier: Essendant
Innovera®
IVR7553X IVR7553A
500037-482EA 70.79 USD InStock
500037-482 500028-957
Innovera® Laser Cartridge, 7553A, 7553MICR, 7553X, Essendant LLC MS
Inks
Designed for fast reliable printing.

  • Cartridge with 5% coverage
  • Compatible and remanufactured

Will handle the toughest workloads with ease. Simple to install for error-free loading.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty