Would you like to log in again?
Crews® ForceFlex™ Safety Glasses
Supplier: MCR Safety
Crews®
CRWFF122 CRWFF128B CRWFF120
500012-794EA 15.83 USD InStock
500012-794 500012-785 500012-790
Crews® ForceFlex™ Safety Glasses
Safety Glasses
Flexible frame combines with lenses to achieve military-level impact resistance.
  • Scratch-resistant
  • Exceeds ANSI Z87.1+
  • Stylish and comfortable
Thermoplastic frames flex to fit any size face. Meets military test regiment for high-velocity impact protection.
Certifications: Exceeds ANSI Z87.1+ standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty