Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Narrow Profile Bookcase With Finished Back, Essendant LLC MS

Supplier: Janitorial Supplies
Alera®
ALEBCS67212MY ALEBCS67212MC
500002-777EA 278.63 USD InStock
500002-777 500002-776
Alera® Narrow Profile Bookcase With Finished Back, Essendant LLC MS
Book Holders
Space-saving stylish bookcase with five 11.75" deep shelves. Each shelf accommodates three-ring binders and large publications.

  • Quick-lock fasteners allow easy assembly
  • Square corner/edge style
  • Shelves are 1" thick and adjust in 1.25" increments
  • UV coated
  • Shelf Capacity (Weight): 90 lbs.

One-piece full-finished Veneer back provides a professional appearance.

Certifications: GREENGUARD Children & Schools Certified, Meets or exceeds ANSI/BIFMA standards.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty