Would you like to log in again?
Peroxidase, VWR®
Supplier: VWR Chemicals
0417-100,000U
97061-626EA 447.56 USD InStock
97061-626
Peroxidase, VWR®
HRP catalyzes the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates.
Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty