Would you like to log in again?
VWR® Freezing Media, Bacterial
Supplier: Avantor
N400-25PK
97063-178EA 159.45 USD InStock
97063-178
VWR® Freezing Media, Bacterial
  • 85% post-thaw viability of frozen cultures
  • Easy-to-label vials
  • 25 x 2 ml cryotubes in convenient storage box
VWR offers high purity, bacterial freezing media. The media contains 0.5 ml of LB broth (Miller) with 30% glycerol per tube. Simply add 1 ml of saturated culture to the vial, vortex, and store at –70°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty