Would you like to log in again?
Formagel® Fixative and Transport System, Prefilled Containers, Azer Scientific
Supplier: Azer Scientific
Formagel®
PFG-20 PFG-40 PFG-90-CA PFG-20 PFG-60 PFG-90 PFG-60 PFG-40
89194-772CS 90.83 USD InStock
89194-772 CA-11010-424 CA-89194-770 89194-768 CA89194-768 CA-89194-772 89239-472 89194-770
Formagel® Fixative and Transport System, Prefilled Containers, Azer Scientific
Sample Containers General Purpose Containers
Fixation gel is designed to safely and securely transport tissue specimens without the hazards of leaking formalin.
  • Fixation Results Identical To Standard 10% Neutral-Buffered Formalin
  • Excellent Staining Results With Routine, Special, And IHC Staining
  • Polypropylene Containers Half Filled With Gel
  • Polypropylene Cap
Made of 10% neutral-buffered formalin, the gel preserves and fixes tissue samples with identical results to standard formalin. Each polypropylene container is half filled with gel and has a polyethylene screw-on cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty