Would you like to log in again?
Arch Punch Sets, General Tools, ORS Nasco
Supplier: ORS Nasco
318-1271ST
300002-290PK 131.79 USD InStock
300002-290
Arch Punch Sets, General Tools, ORS Nasco
Hole Punches
This punches are designed with close tolerances for creating precise holes in gasket material, rubber and soft metals.
  • Heat treated Sheffield forged steel
Delivery information: Delivered in a plastic roll pouch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty